ENA429359942 Enamine

4-(2-fluorophenyl)-1,3-oxazol-2-amine

  • MDL number MFCD16667959
  • Cas Number : 1247986-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189030

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189030
Availability details for
Quantity
  • Availability
  • Add to Cart