ENA429359946 Enamine

[1-(4-fluorophenyl)-2-methylpropyl](methyl)amine

  • MDL number MFCD12179559
  • Cas Number : 1157105-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189041
Availability details for
Quantity
  • Availability
  • Add to Cart