ENA429359947 Enamine

methyl 2-[2-(bromomethyl)phenyl]acetate

  • MDL number MFCD14666530
  • Cas Number : 13737-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189042
Availability details for
Quantity
  • Availability
  • Add to Cart