ENA429359948 Enamine

2-(quinazolin-4-ylsulfanyl)butanoic acid

  • MDL number MFCD06641667
  • Cas Number : 781656-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189043
Availability details for
Quantity
  • Availability
  • Add to Cart