ENA429359949 Enamine

2-amino-6-{[methyl(pyrrolidin-3-yl)amino]methyl}-3,4-dihydropyrimidin-4-one hydrochloride

  • MDL number MFCD28118774
  • Cas Number : 1803570-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189044
Availability details for
Quantity
  • Availability
  • Add to Cart