ENA429359950 Enamine

methyl 3-(4-cyanophenyl)prop-2-ynoate

  • MDL number MFCD13191992
  • Cas Number : 1032041-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189047

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189047
Availability details for
Quantity
  • Availability
  • Add to Cart