ENA429359953 Enamine

1-[4-(2-amino-4-fluorophenyl)-1,4-diazepan-1-yl]ethan-1-one

  • MDL number MFCD16271204
  • Cas Number : 1275755-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189053
Availability details for
Quantity
  • Availability
  • Add to Cart