ENA429359969 Enamine

1-azaspiro[4.4]nonane hydrochloride

  • MDL number MFCD22421990
  • Cas Number : 1417567-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189071
Availability details for
Quantity
  • Availability
  • Add to Cart