ENA429359970 Enamine

1-(2-fluorophenyl)-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD17189513
  • Cas Number : 1244979-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189074
Availability details for
Quantity
  • Availability
  • Add to Cart