ENA429359971 Enamine

3-fluoro-4-[(4-methyl-1,3-thiazol-2-yl)oxy]aniline dihydrochloride

  • MDL number MFCD28118778
  • Cas Number : 1795276-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189079
Availability details for
Quantity
  • Availability
  • Add to Cart