ENA429359982 Enamine

4-(1-ethyl-1H-pyrazol-5-yl)piperidine

  • MDL number MFCD14707675
  • Cas Number : 442876-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189091
Availability details for
Quantity
  • Availability
  • Add to Cart