ENA429359989 Enamine

1-[(2-methoxyphenyl)methyl]piperidin-4-amine dihydrochloride

  • MDL number MFCD13461785
  • Cas Number : 57645-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189103

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189103
Availability details for
Quantity
  • Availability
  • Add to Cart