ENA429360002 Enamine

3-(trifluoromethyl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD20693367
  • Cas Number : 35581-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189138

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189138
Availability details for
Quantity
  • Availability
  • Add to Cart