ENA429360003 Enamine

2-[(pyridin-2-yl)methyl]aniline

  • MDL number MFCD21234598
  • Cas Number : 53114-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-70
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189139
Availability details for
Quantity
  • Availability
  • Add to Cart