ENA429360004 Enamine

2-amino-3-methyl-1-phenylbutan-1-ol

  • MDL number MFCD11934853
  • Cas Number : 67323-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189142
Availability details for
Quantity
  • Availability
  • Add to Cart