ENA429360007 Enamine

3-fluoro-4-(trifluoromethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD13185645
  • Cas Number : 694472-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189148
Availability details for
Quantity
  • Availability
  • Add to Cart