ENA429360008 Enamine

4-bromo-5-phenyl-1,2-oxazol-3-amine

  • MDL number MFCD00228032
  • Cas Number : 62847-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189149

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189149
Availability details for
Quantity
  • Availability
  • Add to Cart