ENA429360010 Enamine

3-(1,3-thiazol-2-yl)phenol

  • MDL number MFCD14529354
  • Cas Number : 35582-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189151
Availability details for
Quantity
  • Availability
  • Add to Cart