ENA429360011 Enamine

6-chloro-N-methylpyridin-3-amine

  • MDL number MFCD10697831
  • Cas Number : 120739-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189152
Availability details for
Quantity
  • Availability
  • Add to Cart