ENA429360016 Enamine

N-(piperidin-3-yl)-4-(trifluoromethyl)benzamide hydrochloride

  • MDL number MFCD28118798
  • Cas Number : 1582662-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189166
Availability details for
Quantity
  • Availability
  • Add to Cart