ENA429360023 Enamine

N-methyl-N-(piperidin-4-yl)-4-(trifluoromethyl)benzamide hydrochloride

  • MDL number MFCD28118801
  • Cas Number : 1580795-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189173
Availability details for
Quantity
  • Availability
  • Add to Cart