ENA429360027 Enamine

3,3-dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid

  • MDL number MFCD03426825
  • Cas Number : 133609-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189180
Availability details for
Quantity
  • Availability
  • Add to Cart