ENA429360028 Enamine

1-(2-bromoethyl)-2,3-dichlorobenzene

  • MDL number MFCD12024971
  • Cas Number : 958027-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189181

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189181
Availability details for
Quantity
  • Availability
  • Add to Cart