ENA429360029 Enamine

1-{5-[(1,2-benzoxazol-3-yl)methyl]-1,2,4-oxadiazol-3-yl}ethan-1-amine hydrochloride

  • MDL number MFCD28118609
  • Cas Number : 1795508-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187184
Availability details for
Quantity
  • Availability
  • Add to Cart