ENA429360041 Enamine

methyl 4-amino-2-chloro-3-methoxybenzoate

  • MDL number MFCD23707452
  • Cas Number : 1427452-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189206

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189206
Availability details for
Quantity
  • Availability
  • Add to Cart