ENA429360052 Enamine

4-amino-2,3-dihydro-1H-1,3-benzodiazol-2-one

  • MDL number MFCD18089179
  • Cas Number : 75370-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189225
Availability details for
Quantity
  • Availability
  • Add to Cart