ENA429360053 Enamine

2,4,6-trifluoropyridine-3-carboxylic acid

  • MDL number MFCD18382775
  • Cas Number : 3512-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189226
Availability details for
Quantity
  • Availability
  • Add to Cart