ENA429360054 Enamine

3-bromo-4-cyanobenzene-1-sulfonyl chloride

  • MDL number MFCD18390731
  • Cas Number : 1201684-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189227
Availability details for
Quantity
  • Availability
  • Add to Cart