ENA429360057 Enamine

1-phenyl-1H-1,2,3-triazole-5-carbaldehyde

  • MDL number MFCD22024316
  • Cas Number : 59401-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189231
Availability details for
Quantity
  • Availability
  • Add to Cart