ENA429360059 Enamine

hex-5-en-3-ol

  • MDL number MFCD00021923
  • Cas Number : 688-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189235

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-189235
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart