ENA429360060 Enamine

3-chloro-2-(chloromethyl)thiophene

  • MDL number MFCD16606495
  • Cas Number : 112433-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187186

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187186
Availability details for
Quantity
  • Availability
  • Add to Cart