ENA429360062 Enamine

3-(1-methyl-1H-pyrazol-5-yl)cyclohexan-1-one

  • MDL number MFCD20336548
  • Cas Number : 1339391-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189240
Availability details for
Quantity
  • Availability
  • Add to Cart