ENA429360064 Enamine

4-(difluoromethoxy)-3-fluorobenzene-1-sulfonyl chloride

  • MDL number MFCD12165952
  • Cas Number : 1178111-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189252
Availability details for
Quantity
  • Availability
  • Add to Cart