ENA429360069 Enamine

4-(3-amino-1,2,4-oxadiazol-5-yl)pyridin-2-amine

  • MDL number MFCD21199153
  • Cas Number : 1482849-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189270
Availability details for
Quantity
  • Availability
  • Add to Cart