ENA429360076 Enamine

1-(1-phenylpiperidin-3-yl)methanamine

  • MDL number MFCD20726738
  • Cas Number : 1232059-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189278
Availability details for
Quantity
  • Availability
  • Add to Cart