ENA429360082 Enamine

(2-chloro-4-cyclopropylphenyl)boronic acid

  • MDL number MFCD21099611

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189292

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189292
Availability details for
Quantity
  • Availability
  • Add to Cart