ENA429360090 Enamine

5-amino-2-fluoro-4-methyl-N-phenylbenzene-1-sulfonamide

  • MDL number MFCD19097938
  • Cas Number : 1409737-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189306
Availability details for
Quantity
  • Availability
  • Add to Cart