ENA429360091 Enamine

4-hydrazinyl-6-(methylsulfanyl)pyrimidine

  • MDL number MFCD19201836
  • Cas Number : 99469-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189313
Availability details for
Quantity
  • Availability
  • Add to Cart