ENA429360094 Enamine

5-chloro-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00154760
  • Cas Number : 31217-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189317
Availability details for
Quantity
  • Availability
  • Add to Cart