ENA429360104 Enamine

1-(2-oxopropyl)-5-(trifluoromethyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD18262548
  • Cas Number : 1339334-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189352
Availability details for
Quantity
  • Availability
  • Add to Cart