ENA429360108 Enamine

2-propylcyclopentan-1-one

  • MDL number MFCD04363947
  • Cas Number : 1193-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189380

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189380
Availability details for
Quantity
  • Availability
  • Add to Cart