ENA429360109 Enamine

5-fluoro-2-(piperidin-4-ylmethoxy)pyridine

  • MDL number MFCD16714117
  • Cas Number : 1248781-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189382

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189382
Availability details for
Quantity
  • Availability
  • Add to Cart