ENA429360110 Enamine

6-bromo-3,3-dimethyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD17267475
  • Cas Number : 67159-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189383
Availability details for
Quantity
  • Availability
  • Add to Cart