ENA429360111 Enamine

N-{[3-(trifluoromethyl)phenyl]methyl}prop-2-enamide

  • MDL number MFCD12091167
  • Cas Number : 1155979-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189384

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189384
Availability details for
Quantity
  • Availability
  • Add to Cart