ENA429360112 Enamine

5-bromoimidazo[2,1-b][1,3]thiazole-2-carboxylic acid

  • MDL number MFCD24194455
  • Cas Number : 1550302-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189385

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-189385
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart