ENA429360115 Enamine

3-(2-methoxyethyl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD14661388
  • Cas Number : 1191998-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189388
Availability details for
Quantity
  • Availability
  • Add to Cart