ENA429360117 Enamine

3-(oxan-4-yl)propan-1-ol

  • MDL number MFCD11226124
  • Cas Number : 428871-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189390

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189390
Availability details for
Quantity
  • Availability
  • Add to Cart