ENA429360119 Enamine

4-(3-chloropropoxy)benzaldehyde

  • MDL number MFCD00021077
  • Cas Number : 82625-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187203
Availability details for
Quantity
  • Availability
  • Add to Cart