ENA429360121 Enamine

3-(3-aminophenyl)-5,5-dimethylimidazolidine-2,4-dione

  • MDL number MFCD14608436
  • Cas Number : 1249525-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189404
Availability details for
Quantity
  • Availability
  • Add to Cart