ENA429360126 Enamine

5-(chloromethyl)-1-propyl-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD28118825
  • Cas Number : 1803588-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189424
Availability details for
Quantity
  • Availability
  • Add to Cart